Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015676.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015676.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015676.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015671.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015671.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015671.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015672.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015672.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015672.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015668.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015668.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015668.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015673.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015673.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015673.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015667.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015667.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015667.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015675.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015675.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015675.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015677.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015677.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015677.mm.1 |
TC0X00002453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015679.mm.1 |
NM_008062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015679.mm.1 |
NM_008062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015679.mm.1 |
TC0X00002453.mm.1 |
MTA TC |