Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290740.1 | NP_001277669.1 | 7 - 8 | 1060 | 106 | |
NM_030560.5 | NP_085037.2 | 8 - 9 | 1118 | 106 | ||
NM_172667.2 | NP_766255.1 | 7 - 8 | 1057 | 106 | ||
GenBank mRNA | AK045589.1 | - | 7 - 8 | 889 | 106 | |
AK077961.1 | - | 8 - 9 | 946 | 106 | ||
AK089988.1 | - | 896 | 106 | |||
AK129404.1 | - | 874 | 106 | |||
AK132001.1 | - | 7 - 8 | 1001 | 106 | ||
AK143352.1 | - | 7 - 8 | 890 | 106 | ||
AK149528.1 | - | 7 - 8 | 884 | 106 | ||
AK166487.1 | - | 7 - 8 | 896 | 106 | ||
BC003993.1 | - | 7 - 8 | 870 | 3492146 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |