Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045556.1 | 68 - 69 | 10199 | 87 | ||
GenBank mRNA | AJ888001.1 | - | 66 - 67 | 9792 | 87 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prosaposin | Psap |
AI037048, SGP-1 |
-1.8kb | Anti-sense | Chr. 10 - 60277628 - 60302600 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4632428N05 gene | 4632428N05Rik |
Dies1, PD-1H, VISTA |
41.9kb | Anti-sense | Chr. 10 - 60346851 - 60428381 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015594.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015608.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015615.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015614.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015604.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015607.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015592.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015600.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015606.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015616.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015617.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015619.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015629.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015630.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015593.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015609.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015611.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015620.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015627.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015598.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015598.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015598.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015610.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015613.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015625.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015591.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015596.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015621.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015624.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015626.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015599.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015612.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015618.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015622.mm.1 | TC1000002233.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NM_001252635.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NM_001252635.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NM_023370.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NM_023370.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NR_045556.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | NR_045556.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000015628.mm.1 | TC1000002233.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 377742 | - | - |