Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
949172 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
237310 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140755 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
17363 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313212 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530309 |
BC024659.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530309 |
BC066054.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530309 |
BC094222.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530309 |
XM_001472442.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530309 |
XM_001473940.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268072 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
954466 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
99409 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
420176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401344 |
AF503943.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401344 |
AK045121.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401344 |
AK077038.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401344 |
NM_001081025.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867442 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
220522 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299727 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
666989 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229442 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
288577 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726565 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856520 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
14536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
566451 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
29663 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
87142 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100601 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153647 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P164820 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P164820 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01674017 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06423466 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01674017 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P164820 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07002418 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652555 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07274272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652634 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652628 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07290276 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07002415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652757 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652553 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06808803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652626 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06891162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652547 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652559 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07274283 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652761 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652750 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07002416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652630 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652557 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25652632 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06767879 |
- |
- |