Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100025961.mm.1 |
NM_001271357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025961.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025963.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025963.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025965.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025965.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025965.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025965.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025966.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025966.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025966.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025966.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025968.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025968.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025968.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025968.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025955.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025955.mm.1 |
NM_001271357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025955.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025955.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025955.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025967.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025967.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025967.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025967.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025952.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025952.mm.1 |
NM_001271357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025952.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025952.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025952.mm.1 |
TC1100002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025956.mm.1 |
NM_001271356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025956.mm.1 |
NM_001271357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025956.mm.1 |
NM_146018.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025956.mm.1 |
NR_073164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025956.mm.1 |
TC1100002862.mm.1 |
MTA TC |