Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081493.2 | NP_001074962.1 | 2 - 3 | 295 | 69 | |
NM_013732.7 | NP_038760.3 | 2 - 3 | 334 | 69 | ||
GenBank mRNA | AK134656.1 | - | 2 - 3 | 270 | 69 | |
AK135007.1 | - | 2 - 3 | 267 | 69 | ||
BC056431.1 | - | 2 - 3 | 292 | 6817832 | 69 | |
CJ172339.1 | - | 2 - 3 | 296 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |