Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271411.1 | NP_001258340.1 | 2 - 3 | 599 | 61 | |
NM_001271412.1 | NP_001258341.1 | 2 - 3 | 529 | 61 | ||
NM_001271413.1 | NP_001258342.1 | 2 - 3 | 438 | 61 | ||
NM_001271414.1 | NP_001258343.1 | 2 - 3 | 526 | 61 | ||
NM_028728.3 | NP_083004.1 | 2 - 3 | 456 | 61 | ||
GenBank mRNA | AF361364.1 | - | 2 - 3 | 533 | 61 | |
AK014802.2 | - | 2 - 3 | 457 | 61 | ||
AK048992.1 | - | 2 - 3 | 440 | 61 | ||
AK150635.1 | - | 2 - 3 | 508 | 61 | ||
AK151187.1 | - | 2 - 3 | 508 | 61 | ||
AK151884.1 | - | 2 - 3 | 508 | 61 | ||
AK152262.1 | - | 2 - 3 | 508 | 61 | ||
AK152499.1 | - | 2 - 3 | 508 | 61 | ||
AK152688.1 | - | 2 - 3 | 508 | 61 | ||
AK155106.1 | - | 2 - 3 | 453 | 61 | ||
AK162939.1 | - | 2 - 3 | 510 | 61 | ||
AK169918.1 | - | 2 - 3 | 453 | 61 | ||
AK170098.1 | - | 2 - 3 | 543 | 61 | ||
AK170355.1 | - | 2 - 3 | 506 | 61 | ||
AK170442.1 | - | 2 - 3 | 508 | 61 | ||
AK170545.1 | - | 2 - 3 | 394 | 61 | ||
AK170593.1 | - | 2 - 3 | 464 | 61 | ||
AK170677.1 | - | 2 - 3 | 464 | 61 | ||
AK170811.1 | - | 2 - 3 | 464 | 61 | ||
AK170883.1 | - | 2 - 3 | 507 | 61 | ||
AK171082.1 | - | 2 - 3 | 569 | 61 | ||
AK171086.1 | - | 2 - 3 | 477 | 61 | ||
AK171201.1 | - | 2 - 3 | 438 | 61 | ||
BC150153.1 | - | 2 - 3 | 471 | 61 | ||
BC150488.1 | - | 2 - 3 | 471 | 40141021 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |