Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253738.1 | NP_001240667.1 | 1 - 2 | 346 | 91 | |
NM_001253740.1 | NP_001240669.1 | 1 - 2 | 346 | 91 | ||
NM_001271764.1 | NP_001258693.1 | 1 - 2 | 346 | 91 | ||
NM_001293749.1 | NP_001280678.1 | 1 - 2 | 346 | 91 | ||
GenBank mRNA | AK050665.1 | - | 1 - 2 | 192 | 91 | |
AK088111.1 | - | 1 - 2 | 206 | 91 | ||
AK088902.1 | - | 1 - 2 | 204 | 91 | ||
AK145238.1 | - | 1 - 2 | 203 | 91 | ||
AK145243.1 | - | 1 - 2 | 203 | 91 | ||
AK145804.1 | - | 1 - 2 | 203 | 91 | ||
AK146349.1 | - | 1 - 2 | 176 | 91 | ||
AK146557.1 | - | 1 - 2 | 328 | 91 | ||
AK151441.1 | - | 1 - 2 | 228 | 91 | ||
AK151500.1 | - | 1 - 2 | 203 | 91 | ||
AK151571.1 | - | 1 - 2 | 202 | 91 | ||
AK151578.1 | - | 1 - 2 | 203 | 91 | ||
AK151595.1 | - | 1 - 2 | 175 | 91 | ||
AK151602.1 | - | 1 - 2 | 203 | 91 | ||
AK151991.1 | - | 1 - 2 | 202 | 91 | ||
AK152260.1 | - | 1 - 2 | 202 | 91 | ||
AK152659.1 | - | 1 - 2 | 175 | 91 | ||
AK153031.1 | - | 1 - 2 | 175 | 91 | ||
AK160011.1 | - | 1 - 2 | 228 | 91 | ||
AK164086.1 | - | 1 - 2 | 203 | 91 | ||
AK165428.1 | - | 1 - 2 | 202 | 91 | ||
AK166836.1 | - | 1 - 2 | 190 | 91 | ||
AK167268.1 | - | 1 - 2 | 180 | 91 | ||
AK167534.1 | - | 1 - 2 | 208 | 91 | ||
AK168843.1 | - | 1 - 2 | 171 | 91 | ||
AK169102.1 | - | 1 - 2 | 174 | 91 | ||
AK169245.1 | - | 1 - 2 | 179 | 91 | ||
AK169821.1 | - | 1 - 2 | 220 | 91 | ||
BC055407.1 | - | 3472 | 91 | |||
BC092045.1 | - | 1 - 2 | 179 | 3591372 | 91 | |
CX237327.1 | - | 1 - 2 | 168 | 91 | ||
X53753.1 | - | 1 - 2 | 214 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |