Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-DMa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010386.4 | NP_034516.3 | 2 - 3 | 677 | 98 | |
GenBank mRNA | AK146950.1 | - | 2 - 3 | 678 | 98 | |
AK160460.1 | - | 1 - 2 | 156 | 98 | ||
AK169900.1 | - | 1 - 2 | 108 | 98 | ||
BC001996.1 | - | 1 - 2 | 128 | 3591389 | 98 | |
BC049992.1 | - | 1 - 2 | 90 | 6394795 | 98 | |
KJ650223.1 | - | 1 - 2 | 85 | 98 | ||
U35324.1 | - | 1 - 2 | 85 | 98 | ||
U35325.1 | - | 1 - 2 | 85 | 98 | ||
U35326.1 | - | 1 - 2 | 85 | 98 | ||
U35327.1 | - | 1 - 2 | 85 | 98 | ||
U35328.1 | - | 1 - 2 | 98 | |||
X62742.1 | - | 1 - 2 | 111 | 98 | ||
X90783.1 | - | 1 - 2 | 85 | 98 | ||
X90784.1 | - | 1 - 2 | 85 | 98 | ||
X90785.1 | - | 1 - 2 | 85 | 98 | ||
X90786.1 | - | 1 - 2 | 85 | 98 | ||
X90787.1 | - | 1 - 2 | 85 | 98 | ||
X90788.1 | - | 1 - 2 | 85 | 98 | ||
X90789.1 | - | 1 - 2 | 85 | 98 | ||
X90790.1 | - | 1 - 2 | 85 | 98 | ||
X90791.1 | - | 1 - 2 | 85 | 98 | ||
X90792.1 | - | 1 - 2 | 85 | 98 | ||
X90793.1 | - | 1 - 2 | 85 | 98 | ||
X90794.1 | - | 1 - 2 | 85 | 98 | ||
X90795.1 | - | 1 - 2 | 85 | 98 | ||
X90963.1 | - | 1 - 2 | 85 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |