Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195033.1 | NP_001181962.1 | 10 - 11 | 861 | 102 | |
GenBank mRNA | AK136846.1 | - | 8 - 9 | 2669 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |