Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300013285.mm.1 |
NM_001195075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013285.mm.1 |
NM_001195075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013285.mm.1 |
TC1300002090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013284.mm.1 |
NM_001195075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013284.mm.1 |
NM_001195075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013284.mm.1 |
TC1300002090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013286.mm.1 |
NM_001195075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013286.mm.1 |
NM_001195075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013286.mm.1 |
TC1300002090.mm.1 |
MTA TC |