Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195673.1 | NP_001182602.1 | 2 - 3 | 241 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |