Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251916.1 | NP_001238845.1 | 1 - 2 | 96 | 116 | |
GenBank mRNA | AB306981.1 | - | 1 - 2 | 96 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |