Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253740.1 | NP_001240669.1 | 5 - 6 | 739 | 120 | |
NM_001293748.1 | NP_001280677.2 | 6 - 7 | 722 | 120 | ||
NM_001293749.1 | NP_001280678.1 | 5 - 6 | 739 | 120 | ||
GenBank mRNA | CX237327.1 | - | 5 - 6 | 561 | 120 | |
U04541.1 | - | 6 - 7 | 692 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |