Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301370.1 | NP_001288299.1 | 2 - 3 | 206 | 106 | |
NM_181546.3 | NP_853524.1 | 2 - 3 | 206 | 106 | ||
GenBank mRNA | AB102947.1 | - | 2 - 3 | 129 | 106 | |
AK045901.1 | - | 2 - 3 | 207 | 106 | ||
BC141262.1 | - | 2 - 3 | 148 | 9055874 | 106 | |
BC145602.1 | - | 2 - 3 | 148 | 9054168 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |