Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104357.1 | 1 - 2 | 244 | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pleiomorphic adenoma gene 1 | Plag1 | -454.0 | Anti-sense | Chr. 4 - 3898769 - 3938434 on NCBI Build GRCm38 | |
Downstream |
short chain dehydrogenase/reductase family 16C, member 5 |
Sdr16c5 |
Rdhe2, Scdr9 |
53.1kb | Anti-sense | Chr. 4 - 3995942 - 4019663 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |