Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430503K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108091.1 | 1 - 2 | 33 | 104 | ||
GenBank mRNA | AK020097.1 | - | 1 - 2 | 34 | 104 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
NK2 homeobox 2, opposite strand |
Nkx2-2os |
Gm14112, Nkx2-2as, Nkx2.2AS, OTTMUSG00000015691 |
Sense | Chr. 2 - 147184083 - 147331681 on NCBI Build GRCm38 | |
Downstream | expressed sequence AI646519 | AI646519 |
Aspax1, OTTMUSG00000015701 |
173.8kb | Anti-sense | Chr. 2 - 147361541 - 147362793 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons |