Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110411.1 | 2 - 3 | 265 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | suppressor of variegation 4-20 homolog 2 (Drosophila) | Suv420h2 |
BC024816, KMT5C, Suv4-20h2 |
-4.5kb | Anti-sense | Chr. 7 - 4740092 - 4747514 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 71, member E2 |
Fam71e2 | 4930401F20Rik | 377.0 | Sense | Chr. 7 - 4753226 - 4771361 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |