Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110973.1 | 14 - 15 | 1840 | 94 | ||
GenBank mRNA | AK035897.1 | - | 14 - 15 | 1737 | 94 | |
BC033503.1 | - | 14 - 15 | 1734 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
acidic (leucine-rich) nuclear phosphoprotein 32 family, pseudogene |
Anp32-ps |
Anp32c, Anp32c-ps, Anp32cl, EG448829 |
-8.7kb | Sense | Chr. 11 - 33394905 - 33395397 on NCBI Build GRCm38 |
Downstream |
gamma-aminobutyric acid (GABA) A receptor, pi |
Gabrp | 108.8kb | Sense | Chr. 11 - 33550781 - 33578978 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |