Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027024.3 | NP_081300.1 | 1 | 83 | 98 | |
GenBank mRNA | AY600990.2 | - | 1 | 101 | 98 | |
BC048681.1 | - | 1 | 83 | 6774445 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |