Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128210.1 | - | 1 - 2 | 42 | 56 | |
AF128211.1 | - | 1 - 2 | 42 | 56 | ||
AF128212.1 | - | 1 - 2 | 42 | 56 | ||
AF128213.1 | - | 1 - 2 | 42 | 56 | ||
BC116851.1 | - | 1 - 2 | 46 | 56 | ||
BC116855.1 | - | 1 - 2 | 46 | 56 | ||
M86672.1 | - | 1 - 2 | 144 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |