Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021274.2 | NP_067249.1 | 1 - 2 | 135 | 62 | |
GenBank mRNA | AF227743.1 | - | 1 - 2 | 96 | 62 | |
AK144279.1 | - | 1 - 2 | 126 | 62 | ||
AK150380.1 | - | 1 - 2 | 126 | 62 | ||
AK150765.1 | - | 1 - 2 | 126 | 62 | ||
AK151210.1 | - | 1 - 2 | 126 | 62 | ||
AK151248.1 | - | 1 - 2 | 126 | 62 | ||
AK151415.1 | - | 1 - 2 | 126 | 62 | ||
AK151534.1 | - | 1 - 2 | 126 | 62 | ||
AK152568.1 | - | 1 - 2 | 126 | 62 | ||
AK152814.1 | - | 1 - 2 | 126 | 62 | ||
AK153181.1 | - | 1 - 2 | 126 | 62 | ||
AK157130.1 | - | 1 - 2 | 126 | 62 | ||
AK157139.1 | - | 1 - 2 | 126 | 62 | ||
AK157589.1 | - | 1 - 2 | 126 | 62 | ||
AK157678.1 | - | 1 - 2 | 126 | 62 | ||
AK172540.1 | - | 1 - 2 | 126 | 62 | ||
BC030067.1 | - | 1 - 2 | 102 | 1446589 | 62 | |
BY165450.1 | - | 1 - 2 | 136 | 62 | ||
M33266.1 | - | 1 - 2 | 100 | 62 | ||
M86829.1 | - | 1 - 2 | 125 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |