Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009440.1 | NP_001009440.1 | 9 - 10 | 780 | 79 | |
GenBank mRNA | AF081781.1 | - | 9 - 10 | 780 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |