Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161886.1 | NP_001155358.1 | 10 - 11 | 1620 | 68 | |
XM_012110487.3 | XP_011965877.1 | 9 - 10 | 1774 | 68 | ||
GenBank mRNA | FJ958367.1 | - | 10 - 11 | 1620 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |