Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161886.1 | NP_001155358.1 | 7 - 8 | 1248 | 94 | |
GenBank mRNA | FJ958367.1 | - | 7 - 8 | 1248 | 94 |