Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130155.1 | NP_001123627.1 | 2 - 3 | 176 | 72 | |
GenBank mRNA | AB364648.1 | - | 2 - 3 | 176 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |