Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130155.1 | NP_001123627.1 | 3 - 4 | 315 | 70 | |
GenBank mRNA | AB364648.1 | - | 3 - 4 | 315 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |