Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106372.3 | XP_011961762.1 | 8 - 9 | 1890 | 86 | |
XM_015104819.2 | XP_014960305.1 | 8 - 9 | 1875 | 86 | ||
XM_015104820.2 | XP_014960306.1 | 7 - 8 | 1683 | 86 | ||
XM_015104821.2 | XP_014960307.1 | 8 - 9 | 1875 | 86 | ||
XM_015104822.2 | XP_014960308.1 | 8 - 9 | 1815 | 86 | ||
XM_015104824.2 | XP_014960310.1 | 8 - 9 | 1875 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |