Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094556.2 | XP_014950042.1 | 2 - 3 | 862 | 98 | |
GenBank mRNA | AF434668.1 | - | 2 - 3 | 482 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |