Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002166.3 | XP_004002215.1 | 3 - 4 | 489 | 67 | |
XM_004002167.4 | XP_004002216.1 | 3 - 4 | 902 | 67 |