Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977187.1 | XP_027832988.1 | 3 - 4 | 424 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |