Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162550.1 | NP_001156022.1 | 2 - 3 | 185 | 76 | |
XM_012188066.3 | XP_012043456.1 | 3 - 4 | 312 | 76 | ||
XM_012188069.3 | XP_012043459.2 | 3 - 4 | 287 | 76 | ||
GenBank mRNA | FJ969416.1 | - | 2 - 3 | 185 | 76 |