Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959790.1 | XP_027815591.1 | 3 - 4 | 373 | 81 | |
XM_027959792.1 | XP_027815593.1 | 3 - 4 | 717 | 81 | ||
XM_027959793.1 | XP_027815594.1 | 4 - 5 | 468 | 81 |