Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012666.4 | XP_004012715.2 | 5 - 6 | 1353 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |