Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968658.1 | XP_027824459.1 | 2 - 3 | 725 | 73 | |
XM_027968659.1 | XP_027824460.1 | 2 - 3 | 713 | 73 | ||
XM_027968660.1 | XP_027824461.1 | 2 - 3 | 693 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |