Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016366.4 | XP_004016415.3 | 4 - 5 | 382 | 80 | |
XM_012096148.3 | XP_011951538.2 | 4 - 5 | 410 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |