Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306114.1 | NP_001293043.1 | 4 - 5 | 775 | 71 | |
XM_012169456.3 | XP_012024846.2 | 5 - 6 | 950 | 71 | ||
XM_027965734.1 | XP_027821535.1 | 5 - 6 | 972 | 71 | ||
GenBank mRNA | KF055859.1 | - | 4 - 5 | 775 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |