Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094475.2 | XP_014949961.2 | 3 - 4 | 504 | 72 | |
XM_027967026.1 | XP_027822827.1 | 3 - 4 | 324 | 72 |