Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003405.3 | XP_004003454.1 | 2 - 3 | 277 | 102 | |
XM_015092705.2 | XP_014948191.1 | 2 - 3 | 629 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |