Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187218.3 | XP_012042608.1 | 3 - 4 | 298 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |