Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019415.4 | XP_004019464.2 | 5 - 6 | 873 | 94 | |
XM_027960121.1 | XP_027815922.1 | 4 - 5 | 606 | 94 |