Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004211.4 | XP_004004260.3 | 4 - 5 | 744 | 78 | |
XM_027964380.1 | XP_027820181.1 | 3 - 4 | 461 | 78 |