Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013064.4 | XP_004013113.1 | 2 - 3 | 409 | 97 | |
XM_004013065.4 | XP_004013114.1 | 2 - 3 | 361 | 97 | ||
XM_027974279.1 | XP_027830080.1 | 2 - 3 | 250 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |