Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161861.1 | NP_001155333.1 | 2 - 3 | 469 | 126 | |
GenBank mRNA | FJ943969.1 | - | 2 - 3 | 469 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |