Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971717.1 | XP_027827518.1 | 1 - 2 | 229 | 89 | |
XM_027971719.1 | XP_027827520.1 | 1 - 2 | 229 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |