Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17H5orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956782.1 | XP_027812583.1 | 2 - 3 | 402 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |