Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011382.4 | XP_004011431.2 | 2 - 3 | 772 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |