Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015414.3 | XP_004015463.2 | 1 - 2 | 1520 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |