Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012672.4 | XP_004012721.1 | 2 - 3 | 484 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |