Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101117554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020724.3 | XP_004020773.1 | 1 - 2 | 132 | 103 |
Gene Symbol: LOC114110580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961470.1 | XP_027817271.1 | 1 - 2 | 125 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |